missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Aluminum Bronze, CDA 955M, C95510, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Aluminum Bronze, CDA 955M, C95510, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Antimony/Carbon/Lead/Tin/Silicon/Zinc/Manganese/Iron/Nickel/Aluminium/Copper/Nitrogen/Oxygen/Beryllium/Bismuth/Boron/Cadmium/Chromium/Magnesium/Silver/Cobalt/Arsenic/Phosphorus |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Antimony 0.004 %/Carbon 0.0058 %/Lead 0.01 %/Tin 0.025 %/Silicon 0.073 %/Zinc 0.11 %/Manganese 0.69 %/Iron 3.82 %/Nickel 4.77 %/Aluminium 9.76 %/Copper 80.7 %/Nitrogen (0.0001) %/Oxygen (0.0003) %/Beryllium (0.001) %/Bismuth (0.001) %/Boron (0.001) %/Cadmium (0.001) %/Chromium (0.001) %/Magnesium (0.001) %/Silver (0.001) %/Cobalt (0.003) %/Arsenic (0.004) %/Phosphorus (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction