Learn More
Decon™ Contrad 70 Cleaning Agent
Brand:  Decon™ CON705
Additional Details : CAS : 1310-58-3 Weight : 5.25000kg ??product.attributeSet.Transport?? : UN number : 1814 Chem class : 8 Pack group : II
Product Code. 15183306
  • 70.80 € / 5 L
Estimated Shipment: 17-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Contrad 70
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P361-Take off immediately all contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : Potassium hydroxide,Trisodium citrate dihydrate