Learn More
Thermo Scientific™ Contact Plate Applicator
Description
Bring consistency and standardization to your cleanroom surface and personnel monitoring when using contact plates with the Thermo Scientific™ Contact Plate Applicator. Utilizing a stainless steel 500 gram weight and a calibrated built-in precision 10 second timer, the Contact Plate Applicator supports surface sampling in compliance with the ISO standard ISO-18593. The unit can be exposed to a fully validated VHP cycle for use in isolators.
Easy-to-Use
- Simple “one-press” operation
- Spring loaded to accept all major supplier contact plates
- Stainless steel weight can be removed for reduced pressure for vertical and personnel sampling
- Visual and audible timer for calibration and fault alerts
- Ergonomic, permitting best practice roll-on sampling methodology allowing for use on poor surfaces and avoiding trapping air while sampling
- May be exposed to a validated VHP cycle
- Can be cleaned by a range of disinfectants: Sodium Hypochorite, IPA, submersible in sterilizing solutions
- Has a hygienic design with fully encapsulated timer and smooth stainless steel components
- Three year time unit functionality (based on 60 uses per day over a 36 month period) with built-in indicator that alerts the user when the recommended timer unit replacement is due.
Specifications
Specifications
| Format | Accessories |
| Product Type | Contact Plate Applicator |
| Quantity | 1 Unit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.