Learn More
Thermo Scientific™ Contact Plate Applicator
Product Code. p-8382558
Click to view available options
Product Type:
Contact Plate Applicator
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 15625427
Brand: Thermo Scientific™ EM2000A
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Bring consistency and standardization to your cleanroom surface and personnel sampling when using contact plates with the Thermo Scientific™ Contact Plate Applicator.
Bring consistency and standardization to your cleanroom surface and personnel monitoring when using contact plates with the Thermo Scientific™ Contact Plate Applicator. Utilizing a stainless steel 500 gram weight and a calibrated built-in precision 10 second timer, the Contact Plate Applicator supports surface sampling in compliance with the ISO standard ISO-18593. The unit can be exposed to a fully validated VHP cycle for use in isolators.
Be confident in the consistency of your environmental monitoring with the Contact Plate Applicator. Made up of three key parts—(a) plate holder, (b) weight and (c) the timer—the applicator can be operated with the weight attached for surface sampling and without the weight attached for personnel sampling. The Contact Plate Applicator meets ISO 18593 for surface sampling: the total weight is 500g +/- 5g and the timer is 10 sec. +/- 0.1 sec.
Easy-to-Use
  • Simple “one-press” operation
  • Spring loaded to accept all major supplier contact plates
  • Stainless steel weight can be removed for reduced pressure for vertical and personnel sampling
  • Visual and audible timer for calibration and fault alerts
  • Ergonomic, permitting best practice roll-on sampling methodology allowing for use on poor surfaces and avoiding trapping air while sampling
Easy-to-clean
  • May be exposed to a validated VHP cycle
  • Can be cleaned by a range of disinfectants: Sodium Hypochorite, IPA, submersible in sterilizing solutions
  • Has a hygienic design with fully encapsulated timer and smooth stainless steel components
Easy-to-Service
  • Three year time unit functionality (based on 60 uses per day over a 36 month period) with built-in indicator that alerts the user when the recommended timer unit replacement is due. 
Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY
Specifications
Format Accessories
Product Type Contact Plate Applicator
Quantity 1 Unit