Learn More
WTW™ ConOx Ox-Modul Dissolved Oxygen Sensor Module
Click to view available options
Conductivity:
1 to 2 μS/cm
Unit Size:
Each
Specifications
Specifications
| Conductivity | 1 to 2 μS/cm |
| For Use With (Equipment) | ConOx with Instruction Manual |
| Type | Dissolved Oxygen Sensor Module |
| Includes | 3 Exchange membrane heads, cleaning solution, abrasive foil, OxiCal CX calibration vessel, electrolyte and beaker |