missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Connexin 30.3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565923
This item is not returnable.
View return policy
Description
Immunogen sequence: RLYKDYDMPRV VACSVEPCPH TVDCYISR Highest antigen sequence identity to the following orthologs - mouse 86%, rat 90%.
This gene encodes a transmembrane connexin protein that is a component of gap junctions. Mutations in this gene have been associated with erythrokeratodermia variabilis, progressive symmetric erythrokeratoderma and hearing impairment.
Specifications
| Connexin 30.3 | |
| Polyclonal | |
| Unconjugated | |
| GJB4 | |
| Cnx30.3; connexin 30.3; connexin-30.3; CX30.3; Cxn-30.3; EKV; gap junction beta-4 protein; gap junction membrane channel protein beta 4; gap junction protein beta 4; gap junction protein, beta 4; gap junction protein, beta 4, 30.3kDa; Gjb4; Gjb-4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 127534 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NTQ9 | |
| GJB4 | |
| Recombinant Human Connexin 30.3. Recombinant protein control fragment (Product #RP-105008). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction