Learn More
Macherey-Nagel™ Connection Kit
Shop All Macherey-Nagel Products
Click to view available options
For Use With:
CC Column
Unit Size:
Each
Specifications
Specifications
| Type | Connection Kit |
| Packaging Quantity | 1 Kit |
| Includes | 2 Nuts with End Fittings, Two Sleeves and Two Guide Rings |
| For Use With | CC Column |