Learn More
Lenz Laborglasintrumente Conical Joint Clips Type Keck™
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01103712
Additional Details : Weight : 0.60000kg
Description
- For conical ground joints
- Material: POM, outstanding chemical resistance
- Operating temperature up to approx. 150 °C
Specifications
POM | |
For ground joint NS 12/21 | |
Violet |