Learn More
Quickfit™ Single Cone with Long Tip
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
34/35
40/38
43027
45/40
55/44
7/16
Thickness (Metric) Wall:
1mm
1.5mm
1.8mm
2mm
2.3mm
2.5mm
Unit Size:
Pack of 10
Pack of 50
Description
- Useful for construction of stirrer guides
- Unbadged and free from ceramic markings
- Avoids problem of badge charring at high temperatures which can give an untidy appearance
- Long tip cones manufactured with minimum shank length of 90mm
Specifications
Specifications
| Quantity | 10/Pk. |
| Item Description | Dia. Shank: 42mm, Bore: 31mm |
| Type | Single Cone with Long Tip |
| Cone Size | 45/40 |
| Thickness (Metric) Wall | 2.3mm |