Learn More
Conduritol B, TRC
Product Code. 30363921
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30363921 25 mg
30354402 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30363921 Supplier Toronto Research Chemicals Supplier No. TRCC66500025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25348-64-5
Molecular Formula C6 H10 O4
Molecular Weight (g/mol) 146.14
IUPAC Name (1R,2S,3S,4R)-cyclohex-5-ene-1,2,3,4-tetrol
SMILES O[C@@H]1C=C[C@@H](O)[C@H](O)[C@H]1O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C6H10O4/c7-3-1-2-4(8)6(10)5(3)9/h1-10H/t3-,4-,5+,6+/m1/s1
Formula Weight 146.0579
Chemical Name or Material Conduritol B