Learn More
Hanna Instruments™ Conductivity Standard Solution
Product Code. 12796955
Change view
Click to view available options
Boiling Point:
∼100°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Boiling Point Unit Size
12796955 ∼100°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12796955 Supplier Hanna Instruments™ Supplier No. HI7034
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A choice of ready mixed calibration standards traceable to NIST. Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air. Supplied in boxes of 25.
Specifications
Boiling Point ∼100°C
Capacity (Metric) 460 mL
Color Colorless
Type Calibration Standard Solutions
For Use With (Application) For laboratory and industrial use
mV Range 80,000 μS/cm
pH Range ∼7
Volume (Metric) Sample 460 mL
Warranty 5 Years
Water Soluble Yes