Learn More
Hanna Instruments™ Conductivity Standard
Product Code. 10633272
Change view
Click to view available options
Volume (Metric) Sample:
230 mL
Unit Size:
230mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sample Unit Size
10633272 230 mL 230mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10633272 Supplier Hanna Instruments™ Supplier No. HI7031M
  • ONLINE EXCLUSIVE PRICE
    23.68 €
    19.33 € / 230mL
    Save 4.35 €
    18% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Type Conductivity Standard
For Use With (Application) For analytical use
Volume (Metric) Sample 230 mL