Learn More
Motic™ Condenser, Achromatic, Swing-out, BA400/BA410
Product Code. 15415302
Click to view available options
For Use With (Equipment):
BA400 and BA410 Series Upright Compound Microscopes
Unit Size:
Each
Product Code. 15415302
Brand: Motic™ 1101001900631
Only null left
Add to basket
Add to basket
Designed as a swing-out style condenser for added convenience, this achromatic condenser corrects spherical and chromatic aberrations. Motic™ Condenser, Achromatic, Swing-out, BA400/BA410 is suitable for both BA400 and BA410 Upright Compound Microscopes.
  • Swing-out design.
  • Achromatic for spherical and chromatic aberration correction.
  • Includes iris diaphragm.
Specifications
Type Condenser
Description NA: 0.9/0.13
For Use With (Equipment) BA400 and BA410 Series Upright Compound Microscopes
Includes Iris diaphragm