missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C4c Antibody (99-72-18), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-12091MFV450
This item is not returnable.
View return policy
Description
Complement C4c Monoclonal antibody specifically detects Complement C4c in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Complement C4c | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation | |
| 99-72-18 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| basic C4, Basic complement C4, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3, C4B1, C4B12, C4B2, C4B3, C4FMGC164979, CH, Chido form of C4, CO4C4B5, complement C4-B, complement C4B1a, complement component 4B, complement component 4B (Chido blood group), CPAMD3FLJ60561, EC 2.1.1.144, EC 2.7.11 | |
| Synthetic peptide consisting of the last 15C-terminal residues of the C4c a4-domain (Cys-EELVYELNPLDHRGR). | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 721 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction