Learn More
Tenak™ Comfort Rack for Upright Freezers, 75 mm Box Height, with Boxes
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Brand: Tenak™ TE24276B
Additional Details : Weight : 4.90000kg
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
- Include standard white cardboard boxes with 9 x 9 cell dividers
Specifications
75 mm (Box) | |
Upright Freezers | |
4 x 4 |
562 x 140 x 328 mm | |
Boxes (16) | |
Stainless Steel |