Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 25mm Shelf Height
Providing a complete range of racks for microtiter plates in chest freezers. Tenak™ Comfort Racks for Microtiter Plates in Upright Freezers are stackable.
262.00 € - 425.00 €
Specifications
Product Type | Microtiter Plate Rack |
---|---|
Stackable | Yes |
For Use With (Equipment) | Microtiter Plates, Upright Freezers |
No. per Pack | 1 Pack |
Width (Metric) | 135 mm |
Product Code | Brand | Dimensions (L x W x H) | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Holds | Price | Quantity & Availability | ||||
12407080
|
Tenak™
TE35534 |
540 x 135 x 265mm | Plates (60) |
375.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12417080
|
Tenak™
TE35536 |
540 x 135 x 318mm | Plates (72) |
405.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12427100
|
Tenak™
TE35586 |
630 x 135 x 318mm | Plates (84) |
425.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12447050
|
Tenak™
TE35460 |
450 x 135 x 159mm | Plates (30) |
292.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12457050
|
Tenak™
TE35462 |
450 x 135 x 212mm | Plates (40) |
316.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12467030
|
Tenak™
TE35410 |
360 x 135 x 159mm | Plates (24) |
262.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12467050
|
Tenak™
TE35464 |
450 x 135 x 265mm | Plates (50) |
341.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12477030
|
Tenak™
TE35412 |
360 x 135 x 212mm | Plates (32) |
284.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12477050
|
Tenak™
TE35466 |
450 x 135 x 318mm | Plates (60) |
364.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12487030
|
Tenak™
TE35414 |
360 x 135 x 265mm | Plates (40) |
314.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12487050
|
Tenak™
TE35468 |
450 x 135 x 371mm | Plates (70) |
384.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12487070
|
Tenak™
TE35530 |
540 x 135 x 159mm | Plates (36) |
314.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12497030
|
Tenak™
TE35416 |
360 x 135 x 318mm | Plates (48) |
324.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12497050
|
Tenak™
TE35470 |
450 x 135 x 398mm | Plates (75) |
395.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
12497070
|
Tenak™
TE35532 |
540 x 135 x 212mm | Plates (48) |
344.00 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
12497090
|
Tenak™
TE35580 |
630 x 135 x 159mm | Plates (42) |
333.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
Description
- Racks can either be used as one plate per shelf or the plates can be stacked
- With an alert preventing the shelves from sliding out
- External footprint is 135 x 92mm
Specifications
Microtiter Plate Rack | |
Microtiter Plates, Upright Freezers | |
135 mm |
Yes | |
1 Pack | |
25 mm |