Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 2mm Thick
Shop All Fisherbrand Products
Click to view available options
Volume (Metric) Well:
40 μL
50 μL
70 μL
80 μL
100 μL
120 μL
200 μL
1300 μL
Unit Size:
Each
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations |
| For Use With (Equipment) | OmniPAGE Mini Vertical Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 12 |
| Quantity | 1 Each |
| Thickness (Metric) | 2 mm |
| Volume (Metric) Well | 70 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion