Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 2mm Thick
Product Code. 11827593 Shop All Fisherbrand Products
Click to view available options
Volume (Metric) Well:
40 μL
50 μL
70 μL
80 μL
100 μL
120 μL
200 μL
1300 μL
Unit Size:
Each
Product Code. 11827593
Please to purchase this item. Need a web account? Register with us today!
For use in preparatory electrophoresis and high-throughput screening.
  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Recommended for maximising sample volume
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Color Blue
For Use With (Application) Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations
For Use With (Equipment) OmniPAGE Mini Vertical Gel Electrophoresis Unit
Type Comb
No. of Wells 1 Prep, 1 Marker
Quantity 1 Each
Thickness (Metric) 2 mm
Volume (Metric) Well 1300 μL