Learn More
Combretastatin A4, MedChemExpress
Product Code. 16433946
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. 16433946
Brand: Medchem Express HYN2146
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Combretastatin A4 is a microtubule-targeting agent that binds β-tubulin with Kd of 0.4 μM.
Combretastatin A4 is a microtubule-targeting agent that binds β-tubulin with Kd of 0.4 μM.
Specifications
Chemical Name or Material Combretastatin A4
CAS 117048-59-6
Color White
Purity Grade Notes Research
Molecular Formula C18H20O5
Quantity 25 mg
Synonym CRC 87-09
Solubility Information DMSO : 100 mg/mL (316.11 mM; Need ultrasonic)
SMILES OC1=CC(/C=C\C2=CC(OC)=C(OC)C(OC)=C2)=CC=C1OC
Molecular Weight (g/mol) 316.35
Formula Weight 316.35
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death