Learn More
WTW™ Combined Chloride Selective Electrode
The combined electrodes type 800 are excellent in their handling and offer the additional opportunity to also measure small sample volumes, as the ion selective and the reference electrodes are combined in a compact rod. Furthermore, they are convincing with their exceptionally great price/performance ratio.
Brand: WTW™ 106660
Additional Details : Gewicht : 0.25000kg
Specifications
1m | |
Cl 800 BNC | |
Combined Chloride Selective Electrode |