Learn More
B Braun™ Combi-Stopper Closing Cones
Product Code. 10520053
Change view
Click to view available options
Color:
Blue
Red
White
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10520053 White Pack of 100
10712393 Blue Pack of 100
10500053 Red Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10520053 Supplier B Braun™ Supplier No. 4495209
  • ONLINE EXCLUSIVE PRICE
    241.29 €
    29.25 € / Pack of 100
    Save 212.04 €
    87% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
B Braun™ Combi-Stopper Closing Cones are suitable for sterile closings of IV and syringe assemblies
  • Luer Lock fitting male and female
  • Made of PE (Polyethylene)
  • Sterile closing of all kinds of open IV-accesses in IV-Sets
  • Sterile closing of pre-filled syringes
  • Available in red, blue and white
  • Box of 100 pcs
Specifications
Material Polyethylene
Color White
Product Type Combi Stopper
Connection Type Luer Lock
Sterility Sterile