Learn More
Fisherbrand™ Column Clamp
Shop All Fisherbrand Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
- These sturdy, easily adjusted, multi-purpose clamps are ideal for holding large cylindrical glassware and similar objects
- A large, flat thumbscrew/worm-drive permits maximum tightening or removing in minimum time
- Constructed of corrosion-resistant stainless steel.
- Ideal for chromatography columns
Specifications
Specifications
| Product Type | Column clamp |
| Material | Stainless Steel |
| Length (Metric) | 282 mm |
| For Use With (Equipment) | Large cylindrical glassware and similar objects. Ideal for chromatography columns. |