Learn More
Fisherbrand™ Column Clamp
Product Code. 15878994 Shop All Fisherbrand Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 15878994
Only null left
Add to basket
Add to basket
Fisherbrand™ Column Clamp is ideal for holding large cylindrical glassware and similar objects.
  • These sturdy, easily adjusted, multi-purpose clamps are ideal for holding large cylindrical glassware and similar objects
  • A large, flat thumbscrew/worm-drive permits maximum tightening or removing in minimum time
  • Constructed of corrosion-resistant stainless steel.
  • Ideal for chromatography columns
TRUSTED_SUSTAINABILITY
Specifications
Product Type Column clamp
Material Stainless Steel
Length (Metric) 282 mm
For Use With (Equipment) Large cylindrical glassware and similar objects. Ideal for chromatography columns.