Learn More
Columbianadin, MedChemExpress
Product Code. 16494817
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16494817
Brand: Medchem Express HYN0362
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Columbianadin, a natural coumarin from, is known to have various biological activities including anti-inflammatory and anti-cancer effects.
Columbianadin, a natural coumarin from, is known to have various biological activities including anti-inflammatory and anti-cancer effects.
Specifications
Chemical Name or Material Columbianadin
CAS 5058-13-9
Color White
Purity Grade Notes Research
Molecular Formula C19H20O5
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (101.50 mM; Need ultrasonic)
SMILES C/C=C(C)\C(OC(C)([C@@H]1CC2=C(O3)C(C=CC3=O)=CC=C2O1)C)=O
Molecular Weight (g/mol) 328.36
Formula Weight 328.36
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death