Learn More
CPC™ Autoclavable Sanitary SIP Connectors
Click to view available options
Hose Barb:
1/2 Hose Barb
3/8 Hose Barb
Tubing Size:
3/4 x 1-1/2 in.
3/4 x 3/4 in.
Unit Size:
Each
Pack of 5
Specifications
Specifications
| Certifications/Compliance | USP Class VI |
| For Use With (Application) | SIP Processing |
| Tubing Size | 3/4 x 3/4 in. |
| Hose Barb | 3/8 Hose Barb |
| Material (Seal) | Platinum-cured Silicone |
| Format | Free-floating |
| Product Type | ADCF SIP Connector |