missing translation for 'onlineSavingsMsg'
Learn More

COL5A2 Monoclonal Antibody (OTI2H3), TrueMAB™, OriGene

Product Code. 17502024
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17502024 30 μL 30µL
17582014 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17502024 Supplier Origene Technologies Supplier No. TA809611S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COL5A2 Monoclonal antibody specifically detects COL5A2 in Human samples. It is validated for Western Blot

Specifications

Antigen COL5A2
Applications Western Blot
Classification Monoclonal
Clone OTI2H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Pgc
Gene Accession No. P20142
Gene Alias 1110014L14Rik; AB collagen; col5a2; Collagen alpha-2(V) chain; collagen type V alpha 2 chain; collagen, fetal membrane, A polypeptide; collagen, type V, alpha 2; D230017N05; EDSC; procollagen, type V, alpha 2; type V preprocollagen alpha 2 chain
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 17-388 of human PGII produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5225
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.