missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15939
This item is not returnable.
View return policy
Description
Coilin Polyclonal specifically detects Coilin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Coilin | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| CLN80p80-coilin, coilin, coilin p80, p80 | |
| Rabbit | |
| 63 kDa | |
| 0.1 mL | |
| Neuroscience | |
| 8161 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P38432 | |
| COIL | |
| Recombinant protein encompassing a sequence within the center region of human Coilin. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction