Learn More
COD Reagent, ISO, Chem-Lab
Product Code. 11909192
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11909192 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11909192 Supplier Chem-Lab Supplier No. CL02.0316.1000
  • ONLINE EXCLUSIVE PRICE
    106.24 €
    86.00 € / 1 L
    Save 20.24 €
    19% Off
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Determine the amount of pollutant in a water sample. COD Reagent, ISO, Chem-Lab is an oxidizing agent for COD analysis, comprised of mercury sulfate with potassium dichromate in sulfuric acid. The solution meets ISO standards for analysis of water quality.
Chemical oxygen demand (COD) is a measure of the amount of oxygen that can be consumed by reactions in a measured solution. A COD test quantifies the amount of organic materials in waste water or surface water. COD measurements are commonly made on samples to determine the amount of oxidizable pollutants and to determine the effect a particular effluent will have on a receiving body.
  • According to ISO standards
  • 80g HgSO4 + 12.259g K2Cr2O7 per 1L 10% H2SO4
    • Specifications
      Certifications/Compliance ISO
      Composition 80g HgSO4 + 12.259g K2Cr2O7 / 1L 10% H2SO4 = 0.25 N
      Description COD Reagent
      For Use With (Application) COD water determination
      Quantity 1 L
      compliance-icons compliance-icons compliance-icons
      Product Identifier
      • COD reagent acc. ISO
      Signal Word
      • Danger
      Hazard Category
      • Acute toxicity Category 3
      • LONG-TERM AQUATIC HAZARD Chronic 1
      • Carcinogenicity Category 1B
      • Substances and mixtures corrosive to metals Category 1
      • Germ cell mutagenicity Category 1
      • Reproductive toxicity Category 1
      • Skin corrosion/irritation Category 1 B
      • Specific target organ toxicity after repeated exposure Category 2
      Hazard Statement
      • H290-May be corrosive to metals.
      • H301-Toxic if swallowed.
      • H314-Causes severe skin burns and eye damage.
      • H340-May cause genetic defects.
      • H350-May cause cancer.
      • H360-May damage fertility or the unborn child.
      • H373-May cause damage to organs.
      • H410-Very toxic to aquatic life with long lasting effects.
      Precautionary Statement
      • P273-Avoid release to the environment.
      • P280-Wear protective gloves/protective clothing/eye protection/face protection.
      • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
      • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
      • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
      • P308+P313-IF exposed or concerned: Get medical advice/attention.
      • P310-Immediately call a POISON CENTER/doctor/.
      Supplemental information
      • MIXTURE LIST-Contain: Mercury(II) sulfate,Potassium dichromate, Sulphuric acid