missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Coconut oil, pure, refined

Product Code. p-2691232 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10169841

Brand: Thermo Scientific Chemicals 365471000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 8001-31-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-31-8
MDL Number MFCD00130817

Specifications

Acidity 0.5mg KOH/g max.
Melting Point 23.0°C to 26.0°C
Color White
Density 0.9030g/mL
Flash Point >200°C
Packaging Glass bottle
Quantity 100 g
Specific Gravity 0.903
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,chloroform and carbon,disulfide
Grade Pure
Physical Form Unctious Mass
Chemical Name or Material Coconut oil, Refined
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.