missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COCH Polyclonal Antibody
GREENER_CHOICE

Product Code. 16550432
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16550432 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16550432 Supplier Invitrogen™ Supplier No. PA597602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COCH Polyclonal Antibody for Western Blot, ELISA

Cochlin is a secreted protein encoded by the coagulation factor C homology (COCH) gene, a cochlear gene. It constitutes 70% of the inner ear proteins and is classified into three glycosylated isoforms: p63s, p44s and p40. Cochlin contains an N-terminal LCCL domain and two von Willebrand factor A-like domains. Mutations in the COCH gene cause DFNA9, an autosomal dominant nonsyndromic auditory and vestibular dysfunction disorder, as a result of either an amino acid deletion in the LCCL domain or missense substitutions. Microfibrillar deposits accumulate in the inner ear of individuals with DFNA9 and these deposits may contain the Cochlin protein. Cochlin is a target antigen for autoimmune sensorineural hearing loss.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COCH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COCH
Gene Accession No. O43405
Gene Alias AW122937; coagulation factor C homolog (Limulus polyphemus); coagulation factor C homolog, cochlin (Limulus polyphemus); Coch; COCH5B2; COCH-5B2; Cochlin; D12H14S564E; DFNA9; PRO294; UNQ257; UNQ257/PRO294
Gene Symbols COCH
Host Species Rabbit
Immunogen Recombinant Human Cochlin protein (20-260AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1690
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.