Learn More
Ekastu Safety™ Cobra Foldy FFP1 Filtering Facepiece
Product Code. 10526920
Change view
Click to view available options
Product Type:
General Purpose Particulate Respirator
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10526920 General Purpose Particulate Respirator Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10526920 Supplier Ekastu Safety™ Supplier No. 419210
  • ONLINE EXCLUSIVE PRICE
    18.15 €
    16.80 € / Pack of 20
    Save 1.35 €
    7% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protects against fine inert particles up to 4.5 times the NPF value. Ekastu Safety™ Cobra Foldy FFP1 Filtering Facepiece has a five-year shelf life in original packaging.
  • Construction of mask in 3D panel design
  • Foam lined comfort nose zone
  • Flexible nose clip
  • With comfortable, cycled harness and unique comfort-harness clip
  • Metal-free band holder-strap
  • Specially designed for wearer of EKASTU spectacles and goggles
  • Space saving due to fold ability
  • Individually packed to avoid contamination during storage
  • Protection against fine inert particles up to 4.5 times the NPF value
  • Protection class DIN EN 149:2001 FFP1 NR
  • In original packaging: 5 years shelf life
  • 20 pieces per pack
    Specifications
    Certifications/Compliance DIN EN 149:2001 FFP1 NR
    Product Type General Purpose Particulate Respirator
    For Use With (Application) Specially designed for wearers of EKASTU spectacles and goggles
    Quantity 20/Pk.
    Style Cycled Harness and Unique Comfort-harness Clip