missing translation for 'onlineSavingsMsg'
Learn More

CNX-774, MedChemExpress

Product Code. 16415928
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16415928

Brand: Medchem Express HY13943

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CNX-774 is an orally active, irreversible and selective BTK inhibitor, with an IC50 of < 1 nM. CNX-774 specifically targets Cysteine 481 of Btk for covalent modification.

CNX-774 is an orally active, irreversible and selective BTK inhibitor, with an IC50 of < 1 nM. CNX-774 specifically targets Cysteine 481 of Btk for covalent modification.

Specifications

Chemical Name or Material CNX-774
CAS 1202759-32-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H22FN7O3
Quantity 10 mg
Solubility Information DMSO : ≥ 45 mg/mL (90.09 mM)
SMILES O=C(NC)C1=NC=CC(OC2=CC=C(NC3=NC=C(F)C(NC4=CC=CC(NC(C=C)=O)=C4)=N3)C=C2)=C1
Molecular Weight (g/mol) 499.5
Formula Weight 499.5
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.