Learn More
CNX-774, MedChemExpress
Product Code. 16488557
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16488557
Brand: Medchem Express HY13943
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CNX-774 is an orally active, irreversible and selective BTK inhibitor, with an IC50 of < 1 nM. CNX-774 specifically targets Cysteine 481 of Btk for covalent modification.
CNX-774 is an orally active, irreversible and selective BTK inhibitor, with an IC50 of < 1 nM. CNX-774 specifically targets Cysteine 481 of Btk for covalent modification.
Specifications
Chemical Name or Material CNX-774
CAS 1202759-32-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H22FN7O3
Quantity 5 mg
Solubility Information DMSO : ≥ 45 mg/mL (90.09 mM)
SMILES O=C(NC)C1=NC=CC(OC2=CC=C(NC3=NC=C(F)C(NC4=CC=CC(NC(C=C)=O)=C4)=N3)C=C2)=C1
Molecular Weight (g/mol) 499.5
Formula Weight 499.5
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease