missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CNTD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15849483
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15849483 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15849483 Supplier Invitrogen™ Supplier No. PA571074

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 83%; Dog: 93%; Human: 100%; Pig: 93%; Rabbit: 86%; Rat: 93%.

CNTD2 gene ontology annotations related to this gene include mitotic cell cycle; positive regulation of cell cycle; positive regulation of cell proliferation; protein phosphorylation; regulation of cyclin-dependent protein serine/threonine kinase activity; regulation of mitotic nuclear division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CNTD2
Gene Accession No. Q9H8S5
Gene Alias CCNP; CNTD2; cyclin N-terminal domain containing 2; cyclin N-terminal domain-containing protein 2; cyclin P; LOW QUALITY PROTEIN: cyclin N-terminal domain-containing protein 2
Gene Symbols CNTD2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human CNTD2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79935
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.