missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CMG1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10200760
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10200760 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10200760 Supplier Invitrogen™ Supplier No. PA19129

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This gene encodes a core intraflagellar transport (IFT) protein which belongs to a multi-protein complex involved in the transport of ciliary proteins along axonemal microtubules. IFT proteins are found at the base of the cilium as well as inside the cilium, where they assemble into long arrays between the ciliary base and tip. This protein, together with intraflagellar transport protein 81, binds and transports tubulin within cilia and is required for ciliogenesis. Naturally occurring mutations in this gene are associated with amyotrophic lateral sclerosis--frontotemporal dementia and Bardet-Biedl Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Ift74
Gene Accession No. Q96LB3
Gene Alias 1700029H06Rik; b2b796Clo; capillary morphogenesis gene 1 protein; capillary morphogenesis protein 1; CCDC2; CMG1; CMG-1; coiled-coil domain containing 2; coiled-coil domain-containing protein 2; Ift74; intraflagellar transport 74; intraflagellar transport 74 homolog; intraflagellar transport protein 74 homolog; LOW QUALITY PROTEIN: intraflagellar transport protein 74 homolog
Gene Symbols Ift74
Host Species Goat
Immunogen Synthetic peptide corresponding to residues C-KTIVDALHSTSGN, from the C-terminus of IFT74 (aa 588-600).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80173
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.