missing translation for 'onlineSavingsMsg'
Learn More

CLRN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16865035 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16865035 20 μL 20µL
16855035 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16865035 Supplier Proteintech Supplier No. 266301AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a protein that contains a cytosolic N-terminus, multiple helical transmembrane domains, and an endoplasmic reticulum membrane retention signal, TKGH, in the C-terminus. The encoded protein may be important in development and homeostasis of the inner ear and retina. Mutations within this gene have been associated with Usher syndrome type IIIa. Multiple transcript variants encoding distinct isoforms have been identified for this gene.

Specifications

Antigen CLRN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CLRN1
Gene Accession No. P58418, Q8K445
Gene Alias clarin 1, CLRN1, USH3, USH3A, Usher syndrome type 3 protein
Gene Symbols CLRN1
Host Species Rabbit
Immunogen CLRN1 Fusion Protein Ag24289
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229320, 7401
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.