missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Clostridium difficile GDH Monoclonal Antibody (5196)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547561

Product Code. 17846487

  • 504.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specificity: Specific for glutamate dehydrogenase of C. difficile. Non-reactive with mammalian cells. Best ELISA pairs (capture/conjugate): Clostridium difficile GDH Monoclonal antibody (Product # MA5-47557)/Clostridium difficile GDH Monoclonal antibody (Product # MA5-47561); Clostridium difficile GDH Monoclonal antibody (Product # MA5-47558)/Clostridium difficile GDH Monoclonal antibody (Product # MA5-47561); Clostridium difficile GDH Monoclonal antibody (Product # MA5-47559)/Clostridium difficile GDH Monoclonal antibody (Product # MA5-47561); Clostridium difficile GDH Monoclonal antibody (Product # MA5-47560)/Clostridium difficile GDH Monoclonal antibody (Product # MA5-47561). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

Clostridium difficile produces an NAD-specific glutamate dehydrogenase (GDH), which converts l-glutamate into alpha-ketoglutarate through an irreversible reaction. The enzyme GDH is detected in the stool samples of patients with C. difficile-associated disease and serves as one of the diagnostic tools to detect C. difficile infection (CDI). [PMID: 24145018].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clostridium difficile GDH
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Bacteria
Antibody
IgG2a
ELISA
5196
Unconjugated
GDH
Proprietary sequence.
100 μg
Primary
4°C or -20°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.