Learn More
Clorgyline hydrochloride, MedChemExpress
Product Code. 16452137
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16452137
Brand: Medchem Express HY14197A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) that is used in scientific research; structurally related to Pargyline.
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) that is used in scientific research
Specifications
Chemical Name or Material Clorgyline hydrochloride
CAS 17780-75-5
Color White
Purity Grade Notes Research
Molecular Formula C13H16Cl3NO
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (324.01 mM) ∣H2O : 100 mg/mL (324.01 mM; Need ultrasonic)
SMILES C#CCN(CCCOC1=CC=C(Cl)C=C1Cl)C.[H]Cl
Molecular Weight (g/mol) 308.63
Formula Weight 308.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism