Learn More
Clocapramine Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Clocapramine Dihydrochloride |
| CAS | 28058-62-0 |
| Molecular Formula | C28H39Cl3N4O |
| Quantity | 1 mg |
| Synonym | 1'-[3-(3-Chloro-10,11-dihydro-5H-dibenz[b,f]azepin-5-yl)propyl]-[1,4'-bipiperidine]-4'-carboxamide Dihydrochloride, 5H-Dibenz[b,f]azepine [1,4'-Bipiperidine]-4'-carboxamide derivative, 3-Chloro-5-[3-(4-piperidino-4-carbamoylpiperidino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine Dihydrochloride, Clofekton, Y 4153 |
| InChI Formula | InChI=1S/C28H37ClN4O.2ClH/c29-24-12-11-23-10-9-22-7-2-3-8-25(22)33(26(23)21-24)18-6-15-31-19-13-28(14-20-31,27(30)34)32-16-4-1-5-17-32;;/h2-3,7-8,11-12,21H,1,4-6,9-10,13-20H2,(H2,30,34);2*1H |
| SMILES | O=C(C1(N2CCCCC2)CCN(CCCN3C4=CC(Cl)=CC=C4CCC5=CC=CC=C53)CC1)N |
| IUPAC Name | 1'-(3-(3-chloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)propyl)-[1,4'-bipiperidine]-4'-carboxamide hydrogen chloride |
| Molecular Weight (g/mol) | 553.99 |
| Formula Weight | 552.22 |