missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLMP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16127-1-AP-20UL
This item is not returnable.
View return policy
Description
May be involved in the cell-cell adhesion. May play a role in adipocyte differentiation and development of obesity. Is required for normal small intestine development.Specifications
| CLMP | |
| Polyclonal | |
| Unconjugated | |
| CLMP | |
| ACAM, adipocyte adhesion molecule, ASAM, CLMP, CXADR-like membrane protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 286939, 71566, 79827 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K1G0, Q8R373, Q9H6B4 | |
| Clmp | |
| CLMP Fusion Protein Ag9111 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction