Learn More
Invitrogen™ CLK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596845
Description
Immunogen sequence: CISFELLGLS TFDFLKDNNY LPYPIHQVRH MAFQLCQAVK FLHDNKLTHT DLKPENILFV NSDYELTYNL EKKRDERSVK STAVRVVDFG SATFDHEHHS TIVSTRHYRA PEVILELGWS QPCDVWSIGC IIFEYYVGFT LFQTHDNREH LAMMERILGP IPSRMIRKTR KQKYFYRGRL DWDENTSAGR YVRENCKPLR RYLTSEAEEH HQLFDLIESM LEYEPAKRLT LGEALQHPFF ARLRAEPPNK LWDSSRDISR; Positive Samples: BT-474, HeLa, Mouse brain, Mouse lung; Cellular Location: Nucleus, Nucleus speckle, Nucleus speckle.
CLK2 is a serine/threonine kinase that phosphorylates serine/arginine-rich proteins as related to controlling RNA splicing. It is also thought to link cell cycle progression, apoptosis, and telomere length regulation.
Specifications
| CLK2 | |
| Polyclonal | |
| Unconjugated | |
| CLK2 | |
| AU041688; CDC like kinase 2; CDC-like kinase 2; CLK kinase; Clk2; Clk2-Scamp3 protein; dual specificity protein kinase CLK2; EC 2.7.12.1; hCLK2; kinase CLK2; Protein kinase CLK2; Tu52 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1196, 12748, 365842 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35491, P49760 | |
| CLK2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 239-498 of human CLK2 (NP_0039842). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.