missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLIP4 Monoclonal Antibody (OTI5D10)
GREENER_CHOICE

Product Code. 15926744
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15926744 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15926744 Supplier Invitrogen™ Supplier No. MA527151

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLIP4 Monoclonal Antibody for Western Blot, IHC (P)

CLIP4 includes 4 isoforms of the human protein that are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLIP4
Gene Accession No. Q8N3C7
Gene Alias 1700024K14Rik; 1700074B05Rik; 4833417L20Rik; 5830409B12Rik; CAP-Gly domain containing linker protein family member 4; CAP-GLY domain containing linker protein family, member 4; CAP-Gly domain-containing linker protein 4; Clip4; restin-like 2; restin-like protein 2; RGD1311049; RSNL2
Gene Symbols CLIP4
Host Species Mouse
Immunogen Full length human recombinant protein of CLIP4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79745
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.