Learn More
Nickel Electro™ Clifton™ Stainless Steel Flat Lid
Product Code. 13141303
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
NE1, NE2, NE3 series 14 L Baths
NE1, NE2, NE3 series 8 L Baths
Nickel-Electro Clifton 22L Baths
Nickel-Electro Clifton 2L/ 4L Baths
Nickel-Electro NE4 22/28 L Baths
Nickel-Electro NE4 series 14L Baths
Nickel-Electro NE4 series 8L Baths
missing translation for 'unitSize'
Each
Product Code. 13141303
missing translation for 'mfr': Nickel Electro™ LD14
Only null left
Add to basket
Add to basket
Maintain a stable bath temperature and reduce evaporation with a stainless steel lid. Nickel Electro™ Clifton™ Stainless Steel Flat Lid minimises heat loss and reduces evaporation.
  • Stainless steel
  • Insulated
  • Minimises heat loss
  • Reduces evaporation
  • Assists in maintaining bath temperature
Specifications
For Use With (Equipment) NE1, NE2, NE3 series 14 L Baths
Material Stainless Steel