missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CLEC4D/CLECSF8 Monoclonal antibody specifically detects CLEC4D/CLECSF8 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | CLEC4D/CLECSF8 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 413512 |
| Conjugate | PE-Cyanine5.5 |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | CLEC6, CLEC-6, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL |
| Host Species | Mouse |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human CLEC4D/CLECSF8, Gly52-Asn215, Accession # Q8WXI8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?