missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC2A Polyclonal Antibody

Brand: Thermo Scientific PA5114260
This item is not returnable.
View return policy
Description
Specifications
| CLEC2A | |
| Polyclonal | |
| Unconjugated | |
| CLEC2A | |
| CLEC2A; C-type lectin domain family 2 member A; C-type lectin domain family 2, member A; INPE5792; KACL; Keratinocyte-associated C-type lectin; PILAR; Proliferation-induced lymphocyte-associated receptor; UNQ579; UNQ5792; UNQ5792/PRO19597 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 387836 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6UVW9 | |
| CLEC2A | |
| Synthetic peptide directed towards the C-terminal region of Human CLEC2A (aa 114-163). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction