Learn More
CLEC1A Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809213S
Description
CLEC1A Monoclonal antibody specifically detects CLEC1A in Human samples. It is validated for Western BlotSpecifications
| CLEC1A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P16422 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4072 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1H8 | |
| Unconjugated | |
| EPCAM | |
| 5930406N14Rik; CLEC1; CLEC-1; CLEC1A; C-type lectin domain family 1 member A; C-type lectin domain family 1, member A; C-type lectin-like receptor 1; C-type lectin-like receptor-1; RGD1566110; UNQ569/PRO1131 | |
| Human recombinant protein fragment corresponding to amino acids 24-265 of human EPCAM produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.