Learn More
CLEC1A Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809213
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded protein may play a role in regulating dendritic cell function. Alternative splice variants have been described but their biological nature has not been determined. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.Specifications
| CLEC1A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NC01 | |
| CLEC1A | |
| Full length human recombit protein of human CLEC1A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI1H8 | |
| Unconjugated | |
| CLEC1A | |
| CLEC-1, CLEC1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51267 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.