Learn More
CLEC12B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26077-1-AP-150UL
This item is not returnable.
View return policy
Description
Cell surface receptor that protects target cells against natural killer cell-mediated lysis. Modulates signaling cascades and mediates tyrosine phosphorylation of target MAP kinases.Specifications
| CLEC12B | |
| Polyclonal | |
| Unconjugated | |
| CLEC12B | |
| CLEC12B, Macrophage antigen H, UNQ5782 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 387837, 502900, 71183 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q149M0, Q2HXU8 | |
| CLEC12B | |
| CLEC12B Fusion Protein Ag23396 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |