Description
- Transparent polypropylene
- Wide neck
- Graduated li>Flat bottom
- Compatible with Agilent, CTC, Dionex, Knauer, PerkinElmer, Thermo Scientific, Varian, VWR, Waters
Specifications
Specifications
| Description | Plastic Vials with Graduations |
| Color | Clear |
| Diameter (Metric) Outer | 11.6 mm |
| Material | Polypropylene |
| For Use With (Application) | AA, AAS, Amines, Analysis of pH-Sensitive Samples, Analysis of Polar Compounds, Aqueous Samples, CE, CE/MS, Heavy Metal Analysis, HPLC, IC, PFAS Analysis, Phenols, Proteins |
| Graduated | Yes |
| Base Style | Flat Bottom |
| Volume (Metric) Usable | 1.5 mL |
| For Use With (Equipment) | Agilent, CTC, Dionex, Knauer, PerkinElmer, Thermo Scientific, Varian, VWR, Waters |
| Height (Metric) | 32.0 mm |
| Show More |