Learn More
Cole-Parmer Cleanroom Male Luer to Hose Barb Fittings, Straight Adapter, Natural Nylon
Product Code. 30053731 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Unit Size:
Pack of 10
This item is not returnable. View return policy
Product Code. 30053731
Brand: Cole-Parmer 5011005
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing. RoHS compliant
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 3/32 in.
Material Nylon
Packaging Quantity 10/Pk.
Inlet Male Luer, Hose barb
Color Natural