missing translation for 'onlineSavingsMsg'
Learn More

Phenomenex™ Cleanert™ Septa, 18 mm, No Slit, for Screw Cap Headspace Vials

Product Code. 31741506
Change view
Click to view available options
Color:
Blue, White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
31741506 Blue, White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31741506 Supplier Phenomenex™ Supplier No. AV89100
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Accessories for use with chromatography vials and in various chromatography applications.

  • Brand: Cleanert
  • Technique: Accessory
TRUSTED_SUSTAINABILITY

Specifications

Color Blue, White
Diameter (Metric) Outer 18 mm
Material PTFE/Silicone
Pre-slit No
Sterility Sterile
Autoclavable Autoclavable
Product Type Septa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.